HAMSTER SERUM (sterile) (Heat Inactivated), Rockland Immunochemicals
Supplier: Rockland Immunochemical
D106-10-0100 D106-10-0500 D106-10-1000
76230-090EA 1908.64 CAD
76230-090 76230-092 76230-094
HAMSTER SERUM (sterile) (Heat Inactivated), Rockland Immunochemicals
Sera (Serum)
Heat Inactivated Sterile Serum
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Hamster Serum is used as a supplement to cell culture media. Hamster Serum is also suitable for use as a component of bioassays, immunoassays or enzyme assays. Hamster Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation. Hamster Serum is ideal for investigators in Cancer, Immunology, and Cell Biology research.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal