Anti-Protein A Polyclonal Antibody
Supplier: Rockland Immunochemical
200-901-077 200-901-077S
76235-114EA 575.12 CAD
76235-114 76235-116
Anti-Protein A Polyclonal Antibody
Antibodies
Chicken Anti-Protein A Antibody
  • Purified IgY Fraction
  • ELISA, Western Blot
Protein A is 56 kDA surface protein found in the cell wall of Staphylococcus aureus. Protein A has a high affinity to Human IgG, Mouse IgG and others and is therefore suitable for purification and detection of immunoglobulins.
Anti-PROTEIN A (CHICKEN) Antibody has been tested for use in ELISA and immunoblotting. Although not tested, this antibody may also be useful for neutralization assays, immunohistochemistry, flow Cytometry, immunoprecipitation, immunodiffusion, conjugation and most immunological methods requiring high titer and specificity.