Red Blood Cell Depletion Magnetic Beads, Rockland Immunochemicals
Supplier: Rockland Immunochemical
01-1801-20 01-1801-50
76221-884EA 1055.44 CAD
76221-884 76221-886
Red Blood Cell Depletion Magnetic Beads, Rockland Immunochemicals
Beads Magnetic Beads
Red Blood Cell Depletion Magnetic Beads
  • Separation of Human Red Blood Cells, from whole blood.
  • Remaining white blood cells can be used for downstream assays such as immunoassays, Western blots, FACS, and immunohistochemistry.
  • Bead mean diameter is ~0.5 μm, bead concentration is 20 mg/mL.
Red Blood Cell Depletion Magnetic Beads recognizes human red blood cells. Red Blood Cell Depletion Magnetic Beads is recommended for the depletion of human Red Blood Cells in samples prior to downstream immunological and non-immunological assays and has been testedMore Product Information