Olympic Storage Shelving Systems, Midwest Container Systems LLC TE
Supplier: MIDWEST CONTAINER SYSTEMS LLC TE
J2448-63C J1848-74C J2436-74CXJ243
76253-260EA 1449.95 CAD
76253-260 76253-686 76253-742
Olympic Storage Shelving Systems, Midwest Container Systems LLC TE
Furniture Shelves
Olympic wire shelving made of carbon-steel meets the storage needs. Open construction allows use of maximum storage space of cube. Starter units consists of four shelves and four posts.
  • Can be loaded/unloaded from all sides
  • Wire shelving that can change as quickly as your needs change
  • Posts are grooved at 1" intervals along entire length of post
Open wire design that minimizes dust accumulation and allows a free circulation of air and numbered at 2" increments and double-grooved every 8" for easy indentification.
Certifications: NSF Approved.
Order Now
SPECIFICATIONS
Environmentally Preferable