CP STAT 100M Anti-static Bags and Films, Z-Mar Technology
Supplier: Z-MAR TECHNOLOGY INC. TE
CPSTAT100MR CPSTAT100M1012
76246-386EA 3278.86 CAD
76246-386 76246-496
CP STAT 100M Anti-static Bags and Films, Z-Mar Technology
Bags Sample Bags
These bags made of a strong, abrasion resistant material offering excellent electrostatic shielding properties. The metal-in, five layer construction provides superior protection from punctures and tears.
It is free of primary amines, amides and silicone compounds.