R-100 Rotavapor® Rotary Evaporator System, Büchi
Supplier: Büchi
11100V2122
76266-274EA 23676.41 CAD
76266-274
R-100 Rotavapor® Rotary Evaporator System, Büchi
Evaporators Rotary Evaporators
The R-100 Rotavapor® is an entry level Rotavapor®, meeting the most essential needs in evaporation. The V-100 vacuum pump and F-105 recirculating chiller is included and controlled by the I-100 Interface controller.
  • Economical
  • Solid
  • Expandable
  • Reliable operation
  • Flexibility
The R-100 Rotavapor® has a user-friendly manual-lift mechanism for easy positioning of the evaporation flask in to and out of the water bath. The V-100 vacuum pump and F-105 recirculating chiller is included and is controlled by the I-100 Interface controller. This full system has all of the components needed for efficient evaporation. The patented Combi-clip allows for easy flask handling and vapor duct removal.
Certifications: CE and CSA compliant.
Ordering information: All accessories and connections are included for out of the box usage. Optional safety shield, 1 l evaporation flasks, different joint size glassware, valves for in-house cooling and vacuum are available as accessories (sold separately). Please contact your Avantor representative for more information.