PANSORBIN® Cells, Calbiochem®, EMD MilliporeSigma
Supplier: MilliporeSigma
507858-10GM 507858-1GM 507858-50GM 507858-5GM
CA80057-210EA 2025.27 CAD
CA80057-210 CA80057-212 CA80057-216 CA80057-218
PANSORBIN® Cells, Calbiochem®, EMD MilliporeSigma
Cells
PANSORBIN® Cells are heat-killed, formalin-fixed Staphylococcus aureus cells that have a coat of protein A and have been pickled by the method of Kessler.
Used for mitogenic stimulation of B lyphocytes and for immunoprecipitation.
Storage temperature: 2 to 8 °C
Delivery information: Supplied as a 10% suspension of staphylococcus aureus cells in PBS, 0.1% NaN₃, pH 7.2.
Order Now
SPECIFICATIONS
Environmentally Preferable