Miller® Falcon™ Edge Self-Retracting Lifelines, Honeywell Safety
Supplier: Honeywell Safety
Miller
MP50G-LEK MP30G-LEK MP20G-LEK MP20G-LE MP50G-LE MP30G-LE
76299-782EA 2020.83 CAD
76299-782 76299-780 76299-778 76299-776 76299-774 76299-784
Miller® Falcon™ Edge Self-Retracting Lifelines, Honeywell Safety
Fall Protection Fall Lanyards and Ropes
Designed for sharp edge applications with an edge radius of ≥ 0.005" (0.13 mm), providing protection in applications that require anchoring at foot level.
  • Up to 30% lighter than competitive models for easy installation and transport
  • Reduce worker fatigue and increase productivity
  • Equipped with RFID for efficient asset and inspection tracking
  • Durable design with corrosion-resistant internal component
  • Extended service life lowers cost of ownership
  • Capacity to support user weights up to 420 lbs. in all connections at foot level and above
The versatility you need, and the safety you depend on. Designed for performance in edge applications, Falcon™ Edge SRLs offer versatility, reliability, and enhanced safety. Models available for welding applications include a shock absorber cover that is flame retardant and resistant to weld splatter.
Certifications: Meets OSHA 1926.502, ANSI A10.32-2012, ANSI Z359.14-2014, SRL-LE Class B, and CSA Z259.2.2-17 Class SRL-LE requirements.