DNA (Deoxyribonucleic acid) (from salmon sperm)

Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA

201190 201190
CAST201190EA 350.2 CAD
CAST201190 ST201190
DNA (Deoxyribonucleic acid) (from salmon sperm)
DNA (Deoxyribonucleic acid)

Kit contains salmon sperm DNA in its liquid form.


  • Have pH of 7.5
  • Melting point: 0 °C
  • Boiling point: 100 °C


It can easily soluble in cold water and hot water.

MDL Number: MFCD00130922
CAS Number: 9007-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR