ViscoTip® Syringe Tips
Supplier: Eppendorf
0030089502 0030089502
76263-494EA 323.73 CAD
76263-494 CA76263-494
ViscoTip® Syringe Tips
Pipette Tips
Eppendorf ViscoTip® syringe tips in 'Eppendorf Quality' grade excel in handling viscous liquids with unmatched precision. Ideal for biochemistry and molecular biology, they ensure minimal sample retention and ergonomic efficiency in laboratory tasks.
  • Specialized for liquids with a dynamic viscosity from 200 to 14000 mPa's
  • For dispensing volumes from 100 µl to 10 ml in increments of 10 µl
  • New tip design means significantly lower operation forces are used, speeding up work and reducing energy consumption
  • Automatic tip recognition and volume calculation in Repeater® M4/E3/E3x
  • Free of leachables and slip agents (e.g. slip agents, biocides, plasticisers)
  • Color-coded double ring and continuous volume scale distinguish the ViscoTip® from Combitips® advanced
Easily master the highest viscosity liquids with Repeater®/Multipette® pipettes from Eppendorf: The latest member of the Combitips® advanced dispenser tip family, ViscoTip® syringe-style positive displacement tips are specifically designed and optimized for handling high viscosity liquids up to 14,000 mPa's such as glycerol 99.5%, Tween®, oils, creams, shampoos, nail polish, and even honey. Save time by dispensing up to 100 times with one filling, and with sharply reduced operating forces, dispensing is faster and the Repeater® battery charge lasts longer.
Automatic tip recognition and volume calculation of ViscoTip® dispensing needles by the Repeater® M4, Repeater® E3/E3x, and Multipette® multi-dispenser pipettes makes routine work more efficient.
A range of application notes for various high viscosity liquids provide a framework for recommended working speeds and measurement errors.