2X YT Agar Plates with 50 µg/ml Kanamycin, Teknova
Supplier: TEKNOVA CA
Y4365
CA100219-372PK 780.25 CAD
CA100219-372
2X YT Agar Plates with 50 µg/ml Kanamycin, Teknova
Media for Microbiology Prepared Media for Microbiology
2X YT (Yeast Extract Tryptone) is a nutritionally rich growth medium designed for growth of recombinant strains of Escherichia coli. 2X YT contains twice as much yeast extract and 60% more tryptone than LB.
  • Ambient shipping conditions
  • Refrigerate to store
Composition: 1.6% tryptone, 1.0% yeast extract, 0.5% sodium chloride, 1.5% agar, 50 µg/ml kanamycin.