Chlorine - pH Test Kit, LaMotte
Supplier: LaMotte
6980
66130-153EA 998.97 CAD
66130-153
Chlorine - pH Test Kit, LaMotte
Environmental Test Kits
Chlorine pH test kit is used for testing for free available chlorine, monochloramine, dichloramine, total residual chlorine, and pH using DPD tablets, phenol red tables and the octa-slide 2 comparator.
  • Range and sensitivity (low and high): 0.1 to 1.0 and 1.0 to 6.0 ppm Cl
Applications include industrial water boiling and cooling, water and wastewater.