Violet Red Bile Glucose Agar, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M581-500G M581-2.5KG M581-5KG
CA95022-026EA 1065.5 CAD
CA95022-026 CA89225-916 CA89225-918
Violet Red Bile Glucose Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Violet red bile glucose agar without lactose is used for detection and enumeration of Enterobacteriaceae in raw foods and clinical samples.
  • Appearance: Light yellow to pinkish beige homogeneous free flowing powder
  • Gelling: Firm, comparable with 1.2% agar gel