Skim Milk PCA, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M1623-500G M1623-2.5KG M1623-5KG
CA95021-368EA 1885.41 CAD
CA95021-368 CA89224-448 CA89224-450
Skim Milk PCA, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Skim milk plate count agar is recommended for determining the microbial count in milk and dairy products.
  • Appearance: Cream to yellow homogeneous free flowing powder
  • Gelling: Firm, comparable with 1.05% agar gel
Casein enzymic hydrolysate provides amino acids and other complex nitrogenous substances. Yeast extract supplies vitamin B complex. Addition of skim milk in the medium makes the conditions optimal for microorganisms which grow in milk. A wide range of microorganisms can be cultured and enumerated on this medium.