V.C.N. Supplement, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
FD023-5VL
FD023-5X5VL
CA71005-608PK
681.79
CAD
CA71005-608
CA71005-610
V.C.N. Supplement, HiMedia Laboratories
Media for Microbiology
Additives and Supplements for Microbiology
An antibiotic supplement, recommended for the selective isolation of N. gonorrhoeae and N. meningitidis.
- Store at 2 to 8 °C