Protease (from Streptomyces griseus)

Supplier: MilliporeSigma

537088-10KU 537088-25KU 537088-50KU 537088-5KU
CA80602-612EA 333.42 CAD
CA80602-612 CA80602-614 CA80602-616 CA80602-618
Protease (from Streptomyces griseus)
Enzymes

A non-specific protease liquefies mucins and digests proteins to free amino acids.


  • Can be used to isolate living chondrocytes
  • Most active at neutral pH, but stable over wide range of pH and temperature
  • Lyophilized form

Order Now

SPECIFICATIONS

Enzyme name Protease
Environmentally Preferable
Source Streptomyces griseus
Species
CAS No. 9036-06-0
Storage conditions 2...8 °C
Enzyme activity ≥70000 proteolytic units/g dry weight

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR