Stamped Steel Heavy Support Stands with Rods, United Scientific Supplies
Supplier: United Scientific Supplies
SSB4X6-H-VWR SSB5X8-H-VWR SSB6X9-H-VWR SSB6X1-H-VWR
76293-242EA 32.06 CAD
76293-242 76293-244 76293-246 76293-248
Stamped Steel Heavy Support Stands with Rods, United Scientific Supplies
Retort Stands
Heavy stamped steel bases with black enamel finish feature an added steel plate on the back, which makes them about 50% heavier than regular stamped steel bases.
  • Zinc plated rod
The steel plate also facilitates a sturdier threading of the rod into the base.
Ordering information: Bases and Rods are also available individually.
Delivery information: Includes a removable, zinc-plated steel rod.