Cast Iron Support Stands with Rods, United Scientific Supplies
Supplier: United Scientific Supplies
SSB4X6-CI-VWR SSB5X8-CI-VWR SSB6X9-CI-VWR SSB6X1-CI-VWR
76293-622EA 31.14 CAD
76293-622 76293-624 76293-626 76293-628
Cast Iron Support Stands with Rods, United Scientific Supplies
Retort Stands
Tip-free stability for experiments.
  • Glassware can sit directly on base
  • Rods are zinc-plated steel
Heavy cast iron bases with black enamel finish. Includes a removable zinc-plated steel rod.