Stamped Steel Two-Hole Support Stands with Rods, United Scientific Supplies
Supplier: United Scientific Supplies
SSB4X6-H2-VWR SSB5X8-H2-VWR SSB6X9-H2-VWR SSB6X1-H2-VWR
76293-250EA 31.07 CAD
76293-250 76293-252 76293-254 76293-256
Stamped Steel Two-Hole Support Stands with Rods, United Scientific Supplies
Retort Stands
Tip-free stability for experiments.
  • Features two tapped holes
  • zinc plated rod
Stamped steel bases with black enamel finish features 2 tapped holes. Hole at center of the shorter side is for standard ring stand use. Hole at center of the longer side is for applications such as titrations, specific gravity determinations and more.
Ordering information: Bases and Rods are also available individually.
Delivery information: Includes a removable, zinc-plated steel rod.