Crucibles, Tall Form, Glass, United Scientific Supplies
Supplier: United Scientific Supplies
CQP015-VWR CQP025-VWR CQP050-VWR CQP015-L-VWR CQP025-L-VWR CQP050-L-VWR
76293-530EA 130.06 CAD
76293-530 76293-532 76293-534 76293-536 76293-538 76293-540
Crucibles, Tall Form, Glass, United Scientific Supplies
Crucibles
Tall form clear quartz glass crucibles are ideal for applications that require ashing of samples.
  • Clear quartz
  • Low expansion
Quartz material offers low expansion, dimensional stability and thermal shock resistance. Can withstand a continuous operating temperature of 1050 °C (1922 °F).