Sodium carbonate, anhydrous 99,5-100,5 Acidimetry, EMPROVE® EXPERT NF, JP, Ph. Eur., BP BiopharmaceuticalProduction, SAFC®

Supplier: EMD MILLIPORE – PCS CA EXCEP
Warning

Synonyms: Calcined soda, Carbonic acid disodium salt

1.37014.5000 1.37014.9029
CA1.37014.5000EA 653.33 CAD
CA1.37014.5000 CA1.37014.9029
Sodium carbonate, anhydrous 99,5-100,5 Acidimetry, EMPROVE® EXPERT NF, JP, Ph. Eur., BP BiopharmaceuticalProduction, SAFC®
Sodium carbonate

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit.


Certifications: Meets the specifications of Ph. Eur., BP, JP, NF.

Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600
Melting Pt: 854
Density: 2.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Acidimetric; Calculated on Dried Substance) 99.5 - 100.5 %
Appearance of Solution Passes Test
Identity Passes Test
Chloride (Cl) ≤0.0125%
Sulfate (SO₄) ≤0.025%
Heavy metals (as Pb) ≤0.001%
As (Arsenic) ≤0.00031%
Fe (Iron) ≤0.0050%
Alkali Hydroxides and Alkali Hydrogen Carbonates Passes Test
Residual Solvents (Ph. Eur./USP/ICH) Excluded by Manufacturing Process
Loss on Drying (300 °C) ≤0.5%
Endotoxins ≤2.5 I.U./g
Total Aerobic Microbial Count ≤100 CFU/g
Total Yeast and Mould Count ≤100 CFU/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR