Human Recombinant EG-VEGF (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-07AF-100UG 100-07AF-1MG 100-07AF-20UG 100-07AF-500UG 100-07AF-5UG
76293-988EA 699.04 CAD
76293-988 76293-990 76293-992 76293-994 76293-996
Human Recombinant EG-VEGF (Animal free) (from E. coli)
Proteins and Peptides
Endocrine gland-derived vascular endothelial growth factor (EG-VEGF) is an angiogenic growth factor that is expressed in the ovaries, testis, adrenal, and placental tissues. EG-VEGF has mitogenic, chemoattractive, and antiapoptotic functional roles.
  • High quality
  • Low endotoxin
  • Affordable
EG-VEGF signaling is mediated through binding the G protein-coupled receptors prokineticin receptor 1 (PKR1) and prokineticin receptor 2 (PKR2).  Polycystic ovaries display strong EG-VEGF expression that is associated with increased angiogenesis and cyst formation, which could lead to the formation of polycystic ovary syndrome and infertility.
Caution: For research use only.