Mouse Recombinant IL-2 (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-17AF-100UG 200-17AF-1MG 200-17AF-20UG 200-17AF-500UG 200-17AF-5UG
76293-910EA 738.44 CAD
76293-910 76293-912 76293-914 76293-916 76293-918
Mouse Recombinant IL-2 (Animal free) (from E. coli)
Proteins and Peptides
Interleukin 2 (IL-2) is an immunomodulatory cytokine that is produced by lymphocytes.  IL-2 signals through the IL-2R receptor to induce activated T cell proliferation and promote T cell differentiation. IL-2 also stimulates the proliferation and differentiation of B cells, natural killer cells, monocytes, and macrophages.
  • High Quality
  • Low Endotoxin
  • Affordable
Caution: For research use only.