Human Recombinant IL-15 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-86AF-100UG 100-86AF-10UG 100-86AF-1MG 100-86AF-500UG
76293-906EA 846.03 CAD
76293-906 76293-972 76293-974 76293-976 76293-978
Human Recombinant IL-15 (from E. coli)
Proteins and Peptides
Interleukin 15 (IL-15) is a widely expressed proinflammatory cytokine that is structurally and functionally related to interleukin 2 (IL-2). IL-15 signals through JAK kinases to activate STAT3, STAT5, and STAT6 transcription factors. IL-15 regulates the activation of T cells, neutrophils, and macrophages, and is a stimulatory cytokine promoting dendritic cell function. IL-15 expression is dysregulated in chronic inflammatory disorders, including rheumatoid arthritis, psoriasis, and pulmonary inflammatory diseases. Therefore, IL-15 may serve as an effective therapeutic target, due to the beneficial outcomes of IL-15 neutralization in models of psoriasis and diabetes. Human IL-15 shows activity on mouse cells.
  • High quality
  • Low endotoxin
  • Affordable