Stainless Steel Laboratory Scoop, United Scientific Supplies
Supplier: United Scientific Supplies
LSCSS6-VWR LSHND1-VWR LSCSET-VWR
76293-326EA 4.07 CAD
76293-326 76293-328 76293-330
Stainless Steel Laboratory Scoop, United Scientific Supplies
Scoops
Great quality stainless steel scoop with one pointed end for handling small amounts of chemicals.
  • Length is 6.5"
  • Also available with a 5" long hardwood handle
The stainless steel scoop has a narrow design for inserting into bottles. Perfect for removing small amounts of powdered material.
Useful for portioning out powders from small containers.