3-Prong Heavy-Duty Extension Clamp with Stainless Steel Rod, United Scientific Supplies
Supplier: United Scientific Supplies
CLHD03-VWR CLHD01 CLHD02
76293-362EA 80.53 CAD
76293-362 470339-272 470338-906
3-Prong Heavy-Duty Extension Clamp with Stainless Steel Rod, United Scientific Supplies
Clamps General Purpose Clamps
These extension clamps features heavy-duty electroplated zinc alloy construction and dual prong adjustments. Clamps have a maximum temperature use of 80 °C.
  • PVC coated jaws
  • Stainless steel rod
6.75" extension clamp: The 4" long extension rod is constructed of stainless steel. Prongs open to about 1.75", have a recommended maximum grip size of about 1.5", and feature double keyed spring-type adjustment screws. Prongs are covered with non-slip PVC sleeves and includes fiberglass sleeves.
9" extension clamp: The 5" long extension rod is constructed of stainless steel. Prongs open to about 2.75", have a recommended maximum grip size of about 2.5", and feature double keyed spring-type adjustment screws. Prongs are covered with non-slip PVC sleeve and includes fiberglass sleeves.
10.75" extension clamp: The 5" long extension rod is constructed of stainless steel. Prongs open to about 4", have a recommended maximum grip size of about 3.25", and feature double keyed spring-type adjustment screws. Prongs are covered with non-slip PVC sleeves.