Deluxe Burette Clamps, United Scientific Supplies
Supplier: United Scientific Supplies
CLBURDS-VWR CLBURDD-VWR
76293-392EA 42.27 CAD
76293-392 76293-394
Deluxe Burette Clamps, United Scientific Supplies
Clamps General Purpose Clamps
Deluxe Burette clamps are die casted from aluminum and feature black rubber sleeves for firm gripping of burettes.
  • Rubber sleeves
  • Holds burettes up to 30 mm O.D.
The adjustable clamps hold up to 30 mm O.D. burettes, which are held in place by the pressure created by the springs. For use with rods up to 19 mm diameter.