Burette Clamp, Stainless Steel, United Scientific Supplies
Supplier: United Scientific Supplies
CLBUR1N-VWR CLBUR2N-VWR
76293-396EA 15.27 CAD
76293-396 76293-398
Burette Clamp, Stainless Steel, United Scientific Supplies
Clamps General Purpose Clamps
These clamps are constructed from steel and feature plastic covered spring jaws.
  • Available in single and double versions
These clamps accept burettes up to 35 mm O.D., and can be used with support rods up to 1/2" O.D.