Vigreux Distilling Columns, 24/40 Joint, United Scientific Supplies
Supplier: United Scientific Supplies
CDV200-VWR CDV300-VWR CDV400-VWR CDV500-VWR
76293-458EA 96.77 CAD
76293-458 76293-460 76293-462 76293-464
Vigreux Distilling Columns, 24/40 Joint, United Scientific Supplies
Distillation Components Distillation Columns
Vigreux distilling columns for cooling and condensing.
  • Features downward pointing indentations to improve cooling
  • 24/40 sockets
A Vigreux column is a type of air condenser where the simple tube has been modified to include an abundance of downward-pointing indentations. This dramatically increases the surface area per unit length of the condenser.
Delivery information: Includes a 24/40 connector.