Sapphire Bottle Top Dispenser, United Scientific Supplies
Supplier: United Scientific Supplies
BTSR2.5-VWR BTSR5-VWR BTSR10-VWR BTSR30-VWR BTSR60-VWR BTSR100-VWR
76293-872EA 1101.72 CAD
76293-872 76293-874 76293-876 76293-878 76293-880 76293-882
Sapphire Bottle Top Dispenser, United Scientific Supplies
Bottle Top Dispensers
High-quality laboratory instruments for liquid handling needs.
  • Precise
  • Easy to use
  • Autoclavable
Save time and increase lab productivity using the sapphire bottle top dispenser. This convenient dispenser uses a dual valve technology that allows simple rinse and refills without dismounting the dispenser.
User-friendly and high-performance Sapphire Bottle Top Dispensers operate in four modes: Dilution, Rinsing, Dual Liquid Handling, and Re-circulation. Fully autoclavable at 121 °C, 15psi for 10 to 15 minutes.
Certifications: Calibrated ISO 17025.