Sani-Pro® Polypropylene Tubing, Holland Applied Technologies
Supplier: Holland Applied Technologies
T200-23 T200-33.5 T200-39.5
76267-966EA 842.95 CAD
76267-966 76267-968 76267-970
Sani-Pro® Polypropylene Tubing, Holland Applied Technologies
Tubing Transfer Tubing
Sani-Pro® Polypropylene tubing.
  • Available in natural polypropylene or PVDF materials
  • Utilize the SIB fusion system to connect to pipe ends
  • Full lot traceability