Polyethylene Tubing, Associated Bag
Supplier: Associated Bag
27-6-10 27-6-06
76239-722EA 460.16 CAD
76239-722 76239-762
Polyethylene Tubing, Associated Bag
Tubing Transfer Tubing
Layflat low density polyethylene tubing.
Simply cut to desired length, insert product, and heat seal, staple, or tape closed.
Certifications: Meets FDA and USDA specifications for food contact.