CleanTeam® Knit Nylon/Carbon Fiber Electrostatic Dissipative (ESD) Gloves, Protective Industrial Products
Supplier: PIP
40-6410/L 40-6410/S
76238-110CS 71350.83 CAD
76238-110 76238-114
CleanTeam® Knit Nylon/Carbon Fiber Electrostatic Dissipative (ESD) Gloves, Protective Industrial Products
Gloves
These uncoated gloves made from seamless knit nylon and carbon fiber yarns are suitable for use in circuit board manufacturing, munitions, electronics assembly and aerospace.
  • Launderable and reusable without losing ESD properties
  • Seamless knit gloves provide great dexterity
Electrostatic Dissipative (ESD) fiber yarns blended with low lint nylon which reduces static build-up on the glove surface for improved performance in electronics assembly.