High Speed Cutters, Dremel
Supplier: DREMEL SERVICES INC. TE
DR-190 DR-191
76241-452EA 42.63 CAD
76241-452 76241-454
High Speed Cutters, Dremel
Cutters
A versatile, high-quality cutter made of high-grade steel for use on wood, plastic and soft metals.
  • For use in soft metals, plastics and woods
  • Ideal for use on curved surfaces
  • Use the sides of the cutter for effective cutting
  • Teal in color
  • Shank Ø: 0.125"
These cutters are ideal for shaping, hollowing, grooving, slotting, inlaying and making tapered holes.