ESD Safe Divider Box Containers and Vertical Dividers, Lewis Bins
Supplier: Lewisbins+
DV1650-XL DC3060-XL DV22120-XL DV17120-XL DV1070-XL DV2250-XL
76242-508EA 15.93 CAD
76242-508 76245-914 76246-738 76247-878 76248-762 76277-324
ESD Safe Divider Box Containers and Vertical Dividers, Lewis Bins
Boxes ESD-Safe Boxes
Multi-purpose box containers can be divided into compartments on 1.25'' centers with easy-to-add dividers. These dividable containers are ideal for use in commercial, industrial, electronics and healthcare markets for storage, assembly, fabrication and distribution applications.
  • Strong stacking ridge and innovative ribbed walls ensures high performance
  • Compatible with many kinds of racking, shelving and material handling equipment
  • Anti-static cardholders available for ESD-Safe identification
Dividers are available to maximize efficiency and flexibility. For easy insertion of dividers, NDC containers feature molded-in "no guess" textured divider grid and numbered divider slots. Multiple-height polyethylene dividers are available.
Vertical dividers are available in a variety of heights and are contoured for easy insertion.