Air Safety Valves, Zoro
Supplier: ZORO - CUSTOMER SERVICE TE
ST25-1A125 SF50-1A150
76253-290EA 40.21 CAD
76253-290 76253-292
Air Safety Valves, Zoro
Valves Air Regulation Valves
Pneumatic safety valves allow air to escape into the environment rather than into a piece of equipment.
  • Maximum temperature 250 °F
  • Stainless steel spring
  • Resilient silicone elastomer poppet
  • Vibra seal coated thread
Applied in compressed air safety relief, standard flow.
Certifications: ASME, CRN, National board certified.
Caution: Cancer and reproductive harm