Stainless Steel Utility Tanks with Lid (Stock Pot), United Scientific Supplies
Supplier: United Scientific Supplies
STKPT06-VWR STKPT11-VWR STKPT17-VWR STKPT26-VWR STKPT37-VWR STKPT50-VWR STKPT71-VWR STKPT98-VWR STKPT169-VWR
76293-230EA 347.31 CAD
76293-230 76293-232 76293-234 76293-236 76293-238 76293-520 76293-522 76293-524 76293-526
Stainless Steel Utility Tanks with Lid (Stock Pot), United Scientific Supplies
Trays/Dishes/Pots Laboratory Trays/Dishes
Steel pots for a wide range of laboratory uses.
  • Made from 304 stainless steel
  • Unsealed 18 gauge
Utility tanks (stock pots) are constructed from unsealed, medium gauge 304 stainless steel. Exceptional for use with gas, electric, or even induction style stoves. Tank features heat-resistant handles, and encapsulated-aluminum bottom with three layers for even heat distribution and less energy consumption.
Delivery information: Cover is included.
Order Now
SPECIFICATIONS
Environmentally Preferable