Airfree® Schlenk Filter Funnels, [ST] Outer Joints, Chemglass
Supplier: Chemglass
AF-0542-05 AF-0542-24 AF-0542-01 AF-0542-02 AF-0542-04 AF-0542-20 AF-0542-25
80064-822EA 873.73 CAD
80064-822 80064-832 80064-814 80064-816 80064-820 80064-826 80064-834
Airfree® Schlenk Filter Funnels, [ST] Outer Joints, Chemglass
Fritted filter funnels are available with a choice of coarse or medium porosity fritted disc.
  • Available with either a 2 mm glass stopcock on the sidearm or a 0 - 4 mm Chem-Vac™ Chem-Cap® valve on the sidearm
  • Lower outlet is standard taper joint
Lower outlet is a standard taper inner joint which features a 0 - 4 mm Chem-Vac™ Chem-Cap® valve. Top outer joint is the same size as the lower inner joint. Capacity listed is that above the fritted disc. Available with a choice of a 2 mm high vacuum glass stopcock with a stainless steel constant pressure retaining device, or a 0 - 4 mm Chem-Cap® Airfree® inlet valve.