SureBond™ 2 IP Paint Microscope Slides, Avantik
Supplier: Avantik
SL6349-1 SL6349-2 SL6349-3 SL6349-4 SL6349-5 SL6349-7
76270-514CS 2308.73 CAD
76270-514 76270-516 76270-518 76270-520 76270-522 76270-524
SureBond™ 2 IP Paint Microscope Slides, Avantik
Slides Microscope Slides
SureBond™ microscope slides provide consistent and reliable results.
  • Optimized to ensure consistent barcode scanning
  • High-quality charge ensures excellent adhesion to eliminate tissue loss
  • 100% quality guarantee
These slides are compatible with all major manufacturer automated printers; resulting in superior print quality.
Order Now
SPECIFICATIONS
Environmentally Preferable