SureBond™ Charged Microscope Slides, Avantik
Supplier: Avantik
SL6332-10 SL6332-5 SL6332-6 SL6332-9 SL6332-4 SL6332-1 SL6332-2 SL6332-7 SL6332-8 SL6332-3
76270-462CS 2012.73 CAD
76270-462 76270-470 76270-208 76270-206 76270-204 76270-468 76270-202 76270-466 76270-464 76270-200
SureBond™ Charged Microscope Slides, Avantik
Slides Adhesive Slides
SureBond™ microscope slides provide consistent and reliable results.
  • Optimized to ensure consistent barcode scanning
  • High-quality charge ensures excellent adhesion to eliminate tissue loss
  • 100% quality guarantee
These slides are compatible with all major manufacturer automated printers; resulting in superior print quality.