Ward's LiveSlides®
Supplier: SOUTHERN SCIENCE SUPPLY
LS50BWB
470309-714BX 274.6 CAD
470309-714 470313-232
Ward's LiveSlides®
Slides Prepared Slides
Never before has it been so easy to view microorganisms, including bacteria, fungi, and protists.
  • Available as bulk pack of 50 slides only, or pack of 5 slides with transfer pipettes
  • Uniform depth of viewing well perfect for maintaining a shallow depth of field
  • Partitions help keep specimens in the viewing area
  • Polycarbonate material is safe for indoor or outdoor use
  • Can be used with traditional or digital microscopes
Each LiveSlide® consists of a base slide, and a set of coverslips of different thicknesses. One is very thin (0.13 mm), perfect forMore Product Information