Ammonium sulfate 99.0-100.5% NF, ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
Management of Change (MOC) category = R
BAKR4628-01
BAKR4628-F9
4628-65
CAJT4628-01CS
43495.58
CAD
CAJT4628-01
CAJT4628-F9
JT4628-65
Ammonium sulfate 99.0-100.5% NF, ACS, J.T.Baker®
Ammonium sulphate
- GMP manufactured product
Formula:
(NH₄)₂SO₄ MW: 132.14 g/mol |
MDL Number:
MFCD00003391 CAS Number: 7783-20-2 |
Specification Test Results
GMP Manufactured Product | |
Meets N.F. Requirements | |
Meets ACS Reagent Chemical Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
NF - Assay ((NH₄)₂SO₄) | 99.0 - 100.5 % |
NF - Insoluble Matter | ≤ 0.005 % |
NF - Residue on Ignition | ≤ 0.005 % |
NF - pH of 5% Solution | 5.0 - 6.0 |
NF - Nitrate (NO₃) | ≤ 0.001 % |
NF - Chloride (Cl) | ≤ 5 ppm |
NF - Phosphate (PO₄) | ≤ 5 ppm |
NF - Iron (Fe) | ≤ 5 ppm |
NF - Identification | Passes Test |
NF - Microbial Limit - Total Aerobic Microbial Count (cfu/g) | ≤ 1000 |
NF - Microbial Limit - Total Combined Molds and Yeasts (cfu/g) | ≤ 10 |
Endotoxin Concentration (EU/g) | ≤ 2.5 |
Nickel (Ni) | ≤ 0.7 ppm |
Cobalt (Co) | ≤ 0.09 ppm |
Chromium (Cr) | ≤ 0.75 ppm |
Copper (Cu) | ≤ 0.03 ppm |
Manganese (Mn) | ≤ 3.45 ppm |
Zinc (Zn) (For Information Only) | |
Ni / Zn Ratio (For Information Only) | |
Co / Zn Ratio (For Information Only) | |
Cu / Zn Ratio (For Information Only) | |
ACS - Assay ((NH₄)₂SO₄) (A.C.S.) | ≥ 99.0 % |
ACS - Residue on Ignition | ≤ 0.005 % |
ACS - pH of 5% Solution at 25°C | 5.0 - 6.0 |
ACS - Insoluble Matter | ≤ 0.005 % |
ACS - Chloride (Cl) | ≤ 5 ppm |
ACS - Nitrate (NO₃) | ≤ 0.001 % |
ACS - Phosphate (PO₄) | ≤ 5 ppm |
ACS - Heavy Metals (as Pb) | ≤ 5 ppm |
ACS - Iron (Fe) | ≤ 5 ppm |
Solution Test | Passes Test |
No Class 1, 2 or 3 Residual Solvents are likely to be present by | |
exclusion of the production/purification process. | |
Elemental Impurities (USP (232), EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. | |
Preserve in well closed container. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...