HardyVal™ CSP Medium Complexity Kit, Comprehensive, Hardy Diagnostics

Supplier: Hardy Diagnostics
HVM1 HVM1
CA89408-160EA 2102.49 CAD
CA89408-160 89408-160
HardyVal™ CSP Medium Complexity Kit, Comprehensive, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
Compliance made easy to verify the efficacy of aseptic technique.

  • The product is quality control tested for growth promotion, pH, and sterility

Hardy Diagnostics Medium Complexity Level Kit, Comprehensive is recommended for simulating medium risk level compounding manipulations involving the preparation of IV Bags and verifying aseptic technique. Kit performs one complete aseptic technique verification challenge test for one technician.

Delivery information: Each Kit Contains:
Tryptic Soy Broth (TSB), 1000 ml Dual Port Bag (1 bag)
Sterile 100 ml Dual Port Bags, empty (6 bags)
Sterile 20 ml Serum Vials, empty (6 vials)
Whirl-Pak™ Bag (1 bag)
Results Log Sheet (1 sheet)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR