TSG Series Small Capacity Refrigerators, Thermo Scientific

Supplier: Avantor
TSG505SA TSG505GA
76204-740EA 13468 CAD
76204-740 76204-742 76204-744 76204-746 76204-748
TSG Series Small Capacity Refrigerators, Thermo Scientific
Refrigerators
The Thermo Scientific TSG Series powered by Phononic∞™ solid state refrigeration technology, has been designed to meet the rigorous and demanding needs of healthcare, research and industrial applications where solid performance is required, in support of breastmilk and medication storage, analyzer kits and reagents, and even cell culture media.

  • Solid state refrigeration technology
  • Whisper quiet operation leading to a more comfortable work environment supporting clinical and healthcare needs
  • Increased capacities due to fewer refrigeration components means
More Product Information
Order Now

SPECIFICATIONS

Cat. No. 76204-740 76204-742 76204-744 76204-746 76204-748
Environmentally Preferable Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient
Electrical 120 V, 60 Hz 100 - 240 V, 50/60 Hz 100 - 240 V, 50/60 Hz 100 - 240 V, 50/60 Hz 100 - 240 V, 50/60 Hz
Plug NEMA 5-15 NEMA 5-15 NEMA 6-15 NEMA 5-15 NEMA 6-15
Environmental certification Energy Star; EPA SNAP Compliant Energy Star; EPA SNAP Compliant Energy Star; EPA SNAP Compliant Energy Star; EPA SNAP Compliant Energy Star; EPA SNAP Compliant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR