Ammonium sulfate 6% (w/v) in aqueous solution, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Diammonium sulfateSulphuric acid diammonium saltsulfuric acid diammonium saltDiammonium sulphateMascagnite

Management of Change (MOC) category = NI

BAKR0333-07 0333-07
CAJT0333-7EA 1034.76 CAD
JT0333-7 CAJT0333-7
Ammonium sulfate 6% (w/v) in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Density: 1.72 g/cm³
MDL Number: MFCD00003391
CAS Number: 7783-20-2

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay ((NH₄)₂SO₄) (by formol method) 5.9 - 6.1 %
pH (As Is" basis)" 5.0 - 6.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR