TLC Tank Rack, PTFE-Coated Aluminum, Chemglass
Supplier: Chemglass
CG-1185-20
80060-620EA 654.09 CAD
80060-620
TLC Tank Rack, PTFE-Coated Aluminum, Chemglass
TLC Development Tanks
PTFE-coated aluminum construction.
  • Designed to fit TLC developing tanks
  • Rack holds six 20 cm×20 cm×1/8" or twelve 20 cm×20 cm×1/16" plates