Improved Arsine Generators, Chemglass
Supplier: Chemglass
CG-1090-01 CG-1090-02
80066-200EA 483.37 CAD
80066-200 80066-202
Improved Arsine Generators, Chemglass
Arsine Generators
For arsine analysis using the diethyldithicarbamate (SDDC) colorimetric method.
  • Compact apparatus requires less time to set up
  • Supllied with a 125mL Erlenmeyer flask, scrubber tube, and a size 24 Keck clip
  • Glass beads may be added to the absorber tube to enhance the gas mixing
Design also meets specifications of ACS/USP, EPA, and can be used with APHA Standard 104A. Apparatus is compact, requires less set-up time and eliminates the ball and socket connection normally used. Complete item consists of a 125 ml Erlenmeyer flask and scrubber tube both having 24/40 standard taper joints. Also supplied complete with one size 24 Keck clip.