Cryo FreezeBox and Accessories, Crystal Technology
Supplier: Crystal
CM02 CM04 FZB-CL-04 CL FZB-S1 CF CM03 FZB-CL-02 FZB-CL-01 CM01 FZB-CL-03
76480-452EA 631.08 CAD
76287-342 76480-452 76287-340 76480-454 76287-350 76480-456 76287-348 76287-338 76287-346 76287-344 76480-450
Cryo FreezeBox and Accessories, Crystal Technology
Coolers Benchtop Coolers
Cooling equipment for your samples. Different Ice Cores available to create your own FreezeBox.
  • No need for dry ice
  • Less contamination
  • Easy to use and bench space friendly
Simple to use. Store the Freezebox Ice Core in ultra-low temperature and place desired module inside. Work on your bench easily with no need for dry ice.
Ordering information: Accessories not included.