Ammonia solution strong NF, ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

Management of Change (MOC) category = R

BAKR3226-20 BAKR3226-45
CAJT3226-20EA 5529.21 CAD
CAJT3226-20 CAJT3226-45
Ammonia solution strong NF, ACS, J.T.Baker®
Ammonia solution

Ammonia Solution, Strong; Ammonium Hydroxide

Formula: NH₃
MW: 35.05 g/mol
MDL Number: MFCD00066650
CAS Number: 1336-21-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Identification Passes Test
NF - Nonvolatile Residue ≤0.02 %
NF - Readily Oxidizable Substances Passes Test
NF - Assay (NH₃)(NF) 27.0 - 30.0 %
ACS - Assay (as NH₃) 28.0 - 30.0 %
ACS - Carbon Dioxide (CO₂) ≤0.002 %
ACS - Residue after Ignition ≤0.002 %
ACS - Substances Reducing Permanganate Passes Test
Appearance Passes Test
Arsenic and Antimony (as As) ≤3 ppm
Chloride (Cl) ≤0.5 ppm
Trace Impurities - Heavy Metals ≤0.5 ppm
Nitrate (NO₃) ≤2 ppm
Phosphate (PO₄) ≤2 ppm
Sulfate (SO₄) ≤2 ppm
Trace Impurities - Aluminum (Al) ≤0.300 ppm
Trace Impurities - Boron (B) ≤0.100 ppm
Trace Impurities - Calcium (Ca) ≤0.300 ppm
Trace Impurities - Chromium (Cr) ≤0.200 ppm
Trace Impurities - Copper (Cu) ≤0.10 ppm
Trace Impurities - Gold (Au) ≤0.300 ppm
Trace Impurities - Iron (Fe) ≤0.2 ppm
Trace Impurities - Lead (Pb) ≤0.200 ppm
Trace Impurities - Magnesium (Mg) ≤0.300 ppm
Trace Impurities - Manganese (Mn) ≤0.200 ppm
Trace Impurities - Nickel (Ni) ≤0.1 ppm
Trace Impurities - Potassium (K) ≤0.300 ppm
Trace Impurities - Sodium (Na) ≤2.000 ppm
Trace Impurities - Tin (Sn) ≤0.300 ppm
Trace Impurities - Titanium (Ti) ≤0.300 ppm
Trace Impurities - Zinc (Zn) ≤0.300 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR