Vacuum Filtration Adapters, with PTFE Stopcocks, Chemglass
Supplier: Chemglass
CG-1053-A-01 CG-1053-A-03
33502-684EA 339.69 CAD
33502-684 33502-688
Vacuum Filtration Adapters, with PTFE Stopcocks, Chemglass
Adapters Adapters, Conical Ground Joints
Adapter used for reduced pressure filtrations using plain stem buchner funnels.
  • Equipped with PTFE stopcock
  • Top of adapter is tooled to accept the listed size of Pluro stopper
Stopcock permits control of vacuum source.
Delivery information: Adapters are not supplied with stoppers.